Assay Details
Gene Symbol: NIBAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243219.1 | NP_001230148.1 | 2 - 3 | 358 | 99 | |
GenBank mRNA | AK346027.1 | - | 2 - 3 | 358 | 99 | |
AK347893.1 | - | 2 - 3 | 322 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
