Assay Details
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243345.1 | NP_001230274.1 | 1 - 2 | 64 | 93 | |
XM_013994186.2 | XP_013849640.1 | 125 | 93 | |||
GenBank mRNA | BP171914.1 | - | 1 - 2 | 67 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243345.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
