Assay Details
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243345.1 | NP_001230274.1 | 4 - 5 | 499 | 88 | |
XM_013994186.2 | XP_013849640.1 | 655 | 88 | |||
GenBank mRNA | AJ945945.1 | - | 1 - 2 | 175 | 88 | |
AK231738.1 | - | 3 - 4 | 657 | 88 | ||
AK237333.1 | - | 4 - 5 | 472 | 88 | ||
AK347687.1 | - | 4 - 5 | 465 | 88 | ||
AK350211.1 | - | 3 - 4 | 652 | 88 | ||
BP171914.1 | - | 4 - 5 | 502 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
