Assay Details
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243367.1 | NP_001230296.1 | 3 - 4 | 750 | 70 | |
XM_021085766.1 | XP_020941425.1 | 819 | 70 | |||
XM_021085767.1 | XP_020941426.1 | 542 | 70 | |||
XM_021085768.1 | XP_020941427.1 | 542 | 70 | |||
GenBank mRNA | AK350557.1 | - | 3 - 4 | 750 | 70 | |
AK389589.1 | - | 3 - 4 | 669 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
