Assay Details
Gene Symbol: PPCDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243368.1 | NP_001230297.1 | 3 - 4 | 341 | 95 | |
XM_005666126.3 | XP_005666183.1 | 415 | 95 | |||
XM_013978050.2 | XP_013833504.1 | 419 | 95 | |||
XM_013978053.2 | XP_013833507.1 | 321 | 95 | |||
XM_013978054.2 | XP_013833508.1 | 296 | 95 | |||
GenBank mRNA | AK237382.1 | - | 3 - 4 | 339 | 95 | |
BW979348.1 | - | 3 - 4 | 334 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013978053.2, XM_013978054.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
