Assay Details
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243497.1 | NP_001230426.1 | 1 - 2 | 210 | 109 | |
XM_021098033.1 | XP_020953692.1 | 4342 | 109 | |||
XM_021098034.1 | XP_020953693.1 | 4342 | 109 | |||
XM_021098036.1 | XP_020953695.1 | 4342 | 109 | |||
XM_021098037.1 | XP_020953696.1 | 4342 | 109 | |||
GenBank mRNA | AK345147.1 | - | 2 - 3 | 345 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
