Assay Details
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243528.1 | NP_001230457.1 | 4 - 5 | 839 | 87 | |
XM_021088564.1 | XP_020944223.1 | 1320 | 87 | |||
GenBank mRNA | AK236429.1 | - | 4 - 5 | 575 | 87 | |
AK240283.1 | - | 4 - 5 | 611 | 87 | ||
AK398677.1 | - | 5 - 6 | 1217 | 87 | ||
AK400153.1 | - | 5 - 6 | 1058 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
