Assay Details
Gene Symbol: EEF1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243545.1 | NP_001230474.1 | 3 - 4 | 423 | 116 | |
GenBank mRNA | BG833879.1 | - | 2 - 3 | 329 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
