Assay Details
Gene Symbol: MTRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243580.1 | NP_001230509.1 | 11 - 12 | 1547 | 67 | |
XM_021065101.1 | XP_020920760.1 | 1809 | 67 | |||
XM_021065102.1 | XP_020920761.1 | 1507 | 67 | |||
XM_021065103.1 | XP_020920762.1 | 1742 | 67 | |||
GenBank mRNA | AK343978.1 | - | 11 - 12 | 1517 | 67 | |
DN117879.1 | - | 3 - 4 | 336 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
