Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243595.1 | NP_001230524.1 | 12 - 13 | 1622 | 91 | |
XM_021090951.1 | XP_020946610.1 | 1974 | 91 | |||
XM_021090952.1 | XP_020946611.1 | 1585 | 91 | |||
XM_021090953.1 | XP_020946612.1 | 1565 | 91 | |||
XM_021090954.1 | XP_020946613.1 | 1571 | 91 | |||
GenBank mRNA | AK343638.1 | - | 12 - 13 | 1622 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
