Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243595.1 | NP_001230524.1 | 4 - 5 | 704 | 81 | |
XM_021090951.1 | XP_020946610.1 | 1038 | 81 | |||
XM_021090952.1 | XP_020946611.1 | 649 | 81 | |||
XM_021090953.1 | XP_020946612.1 | 629 | 81 | |||
XM_021090954.1 | XP_020946613.1 | 653 | 81 | |||
GenBank mRNA | AK343638.1 | - | 4 - 5 | 704 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
