Assay Details
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243663.1 | NP_001230592.1 | 4 - 5 | 299 | 102 | |
XM_021072012.1 | XP_020927671.1 | 1699 | 102 | |||
XM_021072013.1 | XP_020927672.1 | 629 | 102 | |||
GenBank mRNA | AK239039.1 | - | 4 - 5 | 299 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
