Assay Details
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243686.1 | NP_001230615.1 | 5 - 6 | 678 | 124 | |
GenBank mRNA | AK350141.1 | - | 5 - 6 | 678 | 124 | |
DN122554.1 | - | 2 - 3 | 659 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
