Assay Details
Gene Symbol: PRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243801.1 | NP_001230730.1 | 5 - 6 | 823 | 91 | |
GenBank mRNA | AK239604.1 | - | 5 - 6 | 823 | 91 | |
AK394777.1 | - | 4 - 5 | 483 | 91 | ||
AY609594.1 | - | 5 - 6 | 782 | 91 | ||
KU705637.1 | - | 5 - 6 | 822 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
