Assay Details
Gene Symbol: CPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243844.1 | NP_001230773.1 | 5 - 6 | 1219 | 77 | |
GenBank mRNA | AK346161.1 | - | 5 - 6 | 1219 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
