Assay Details
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243913.1 | NP_001230842.1 | 1 - 2 | 207 | 72 | |
GenBank mRNA | AK344489.1 | - | 1 - 2 | 208 | 72 | |
AK399604.1 | - | 1 - 2 | 157 | 72 | ||
DT329164.1 | - | 1 - 2 | 157 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
