Assay Details
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244064.1 | NP_001230993.1 | 1 - 2 | 159 | 107 | |
XM_021079635.1 | XP_020935294.1 | 149 | 107 | |||
GenBank mRNA | AK232978.1 | - | 1 - 2 | 163 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
