Assay Details
Gene Symbol: RPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244070.1 | NP_001230999.1 | 4 - 5 | 631 | 92 | |
GenBank mRNA | AK233375.1 | - | 4 - 5 | 378 | 92 | |
AK236665.1 | - | 936 | 92 | |||
AK347721.1 | - | 4 - 5 | 379 | 92 | ||
AK392151.1 | - | 4 - 5 | 436 | 92 | ||
AK392193.1 | - | 4 - 5 | 379 | 92 | ||
CJ013638.1 | - | 4 - 5 | 631 | 92 | ||
EW573166.2 | - | 2 - 3 | 205 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
