Assay Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244106.1 | NP_001231035.1 | 2 - 3 | 540 | 170 | |
XM_005665896.3 | XP_005665953.2 | 292 | 170 | |||
GenBank mRNA | AK231549.1 | - | 2 - 3 | 187 | 170 | |
AK240249.1 | - | 2 - 3 | 469 | 170 | ||
AK349815.1 | - | 2 - 3 | 188 | 170 | ||
AK349976.1 | - | 2 - 3 | 188 | 170 | ||
AK350605.1 | - | 2 - 3 | 473 | 170 | ||
AK397539.1 | - | 2 - 3 | 187 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
