Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244130.1 | NP_001231059.1 | 3 - 4 | 264 | 110 | |
GenBank mRNA | AK230763.1 | - | 3 - 4 | 264 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
