Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244130.1 | NP_001231059.1 | 5 - 6 | 344 | 103 | |
GenBank mRNA | AK230763.1 | - | 5 - 6 | 344 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
