Assay Details
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244347.1 | NP_001231276.1 | 2 - 3 | 393 | 93 | |
XM_021068214.1 | XP_020923873.1 | 760 | 93 | |||
XM_021068215.1 | XP_020923874.1 | 270 | 93 | |||
XM_021068216.1 | XP_020923875.1 | 521 | 93 | |||
XM_021068217.1 | XP_020923876.1 | 760 | 93 | |||
GenBank mRNA | AK236200.1 | - | 2 - 3 | 395 | 93 | |
AK236359.1 | - | 2 - 3 | 393 | 93 | ||
AK240388.1 | - | 2 - 3 | 231 | 93 | ||
AK347431.1 | - | 2 - 3 | 395 | 93 | ||
AK391059.1 | - | 2 - 3 | 468 | 93 | ||
AK398702.1 | - | 2 - 3 | 232 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244347.1, XM_021068214.1, XM_021068215.1, XM_021068216.1, XM_021068217.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
