Assay Details
Gene Symbol: DCP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244358.1 | NP_001231287.1 | 5 - 6 | 520 | 65 | |
XM_021068463.1 | XP_020924122.1 | 874 | 65 | |||
GenBank mRNA | AK230977.1 | - | 5 - 6 | 624 | 65 | |
AK237144.1 | - | 5 - 6 | 539 | 65 | ||
AK238754.1 | - | 5 - 6 | 539 | 65 | ||
AK347683.1 | - | 5 - 6 | 520 | 65 | ||
AK399200.1 | - | 5 - 6 | 554 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
