Assay Details
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244386.1 | NP_001231315.1 | 1 - 2 | 167 | 101 | |
XM_021100359.1 | XP_020956018.1 | 766 | 101 | |||
GenBank mRNA | AK344118.1 | - | 1 - 2 | 162 | 101 | |
AY609657.1 | - | 1 - 2 | 167 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
