Assay Details
Gene Symbol: MRPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244447.1 | NP_001231376.1 | 3 - 4 | 521 | 76 | |
NM_001244448.1 | NP_001231377.1 | 3 - 4 | 398 | 76 | ||
XM_005663256.3 | XP_005663313.1 | 509 | 76 | |||
XM_005663257.2 | XP_005663314.1 | 568 | 76 | |||
GenBank mRNA | AK231970.1 | - | 3 - 4 | 521 | 76 | |
AK233885.1 | - | 3 - 4 | 476 | 67 | ||
AK394983.1 | - | 3 - 4 | 390 | 76 | ||
AY609955.1 | - | 3 - 4 | 463 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
