Assay Details
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244480.1 | NP_001231409.1 | 2 - 3 | 317 | 78 | |
XM_013987550.2 | XP_013843004.1 | 332 | 78 | |||
GenBank mRNA | AK232399.1 | - | 2 - 3 | 303 | 78 | |
AK237996.1 | - | 2 - 3 | 313 | 78 | ||
AK240587.1 | - | 2 - 3 | 296 | 78 | ||
AK390257.1 | - | 2 - 3 | 317 | 78 | ||
EH006389.1 | - | 2 - 3 | 299 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
