Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244581.1 | NP_001231510.1 | 4 - 5 | 821 | 65 | |
XM_021072058.1 | XP_020927717.1 | 509 | 65 | |||
XM_021072059.1 | XP_020927718.1 | 624 | 65 | |||
GenBank mRNA | EW533941.2 | - | 2 - 3 | 226 | 65 | |
FS666888.1 | - | 4 - 5 | 821 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
