Assay Details
Gene Symbol: EIF3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244624.1 | NP_001231553.1 | 4 - 5 | 305 | 84 | |
GenBank mRNA | AK232367.1 | - | 4 - 5 | 305 | 84 | |
AK394444.1 | - | 4 - 5 | 308 | 84 | ||
AK397989.1 | - | 4 - 5 | 295 | 84 | ||
AY610468.1 | - | 3 - 4 | 262 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
