Assay Details
Gene Symbol: EXOSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244661.1 | NP_001231590.1 | 5 - 6 | 658 | 75 | |
XM_021093302.1 | XP_020948961.1 | 806 | 75 | |||
GenBank mRNA | AK233966.1 | - | 5 - 6 | 658 | 75 | |
AK352190.1 | - | 5 - 6 | 554 | 75 | ||
AK390417.1 | - | 5 - 6 | 759 | 75 | ||
AK398880.1 | - | 5 - 6 | 644 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
