Assay Details
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244730.1 | NP_001231659.1 | 3 - 4 | 312 | 80 | |
XM_005653697.3 | XP_005653754.1 | 377 | 80 | |||
XM_005653698.3 | XP_005653755.1 | 2539 | 80 | |||
XM_005653699.3 | XP_005653756.1 | 368 | 80 | |||
XM_005653701.3 | XP_005653758.1 | 407 | 80 | |||
XM_013977422.2 | XP_013832876.1 | 851 | 80 | |||
XM_013977439.2 | XP_013832893.1 | 447 | 80 | |||
XM_013977441.1 | XP_013832895.1 | 457 | 80 | |||
XM_021102035.1 | XP_020957694.1 | 914 | 80 | |||
XM_021102036.1 | XP_020957695.1 | 1109 | 80 | |||
GenBank mRNA | AK238962.1 | - | 3 - 4 | 312 | 80 | |
AK389867.1 | - | 3 - 4 | 283 | 80 | ||
AK398411.1 | - | 1 - 2 | 690 | 80 | ||
AK399711.1 | - | 4 - 5 | 369 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
