Assay Details
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244730.1 | NP_001231659.1 | 7 - 8 | 1021 | 77 | |
XM_005653697.3 | XP_005653754.1 | 1086 | 77 | |||
XM_005653698.3 | XP_005653755.1 | 3248 | 77 | |||
XM_005653699.3 | XP_005653756.1 | 1077 | 77 | |||
XM_005653701.3 | XP_005653758.1 | 1116 | 77 | |||
XM_013977422.2 | XP_013832876.1 | 1560 | 77 | |||
XM_013977439.2 | XP_013832893.1 | 1156 | 77 | |||
XM_013977441.1 | XP_013832895.1 | 1166 | 77 | |||
XM_021102035.1 | XP_020957694.1 | 1623 | 77 | |||
XM_021102036.1 | XP_020957695.1 | 1818 | 77 | |||
GenBank mRNA | AK238962.1 | - | 7 - 8 | 1021 | 77 | |
AK389867.1 | - | 7 - 8 | 992 | 77 | ||
AK398411.1 | - | 5 - 6 | 1399 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
