Assay Details
Gene Symbol: CCDC181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244796.1 | NP_001231725.1 | 5 - 6 | 1520 | 79 | |
XM_005663096.3 | XP_005663153.1 | 1563 | 79 | |||
XM_013996778.2 | XP_013852232.1 | 1521 | 79 | |||
XM_013996780.2 | XP_013852234.1 | 1803 | 79 | |||
XM_013996781.2 | XP_013852235.1 | 1476 | 79 | |||
GenBank mRNA | DN115207.1 | - | 3 - 4 | 402 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
