Assay Details
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244807.1 | NP_001231736.1 | 2 - 3 | 442 | 62 | |
XM_005670201.3 | XP_005670258.1 | 471 | 62 | |||
GenBank mRNA | AK234091.1 | - | 2 - 3 | 442 | 62 | |
AK236454.1 | - | 2 - 3 | 453 | 62 | ||
AY609740.2 | - | 2 - 3 | 395 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
