Assay Details
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244809.1 | NP_001231738.1 | 4 - 5 | 453 | 106 | |
GenBank mRNA | AK344004.1 | - | 4 - 5 | 454 | 106 | |
AK400047.1 | - | 4 - 5 | 465 | 106 | ||
AY610478.1 | - | 4 - 5 | 386 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
