Assay Details
Gene Symbol: MPZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244821.1 | NP_001231750.1 | 2 - 3 | 352 | 67 | |
GenBank mRNA | AK240334.1 | - | 2 - 3 | 410 | 67 | |
CN156840.1 | - | 2 - 3 | 352 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
