Assay Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137619.1 | NP_001131091.1 | 1 - 2 | 91 | 127 | |
GenBank mRNA | AK231991.1 | - | 1 - 2 | 91 | 127 | |
AK344181.1 | - | 1 - 2 | 92 | 127 | ||
AK392702.1 | - | 1 - 2 | 93 | 127 | ||
AK398969.1 | - | 2 - 3 | 200 | 127 | ||
FJ268645.1 | - | 1 - 2 | 91 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001137619.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
