Assay Details
Gene Symbol: ID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244700.1 | NP_001231629.1 | 2 | 921 | 130 | |
GenBank mRNA | AK231961.1 | - | 2 | 927 | 130 | |
AK348412.1 | - | 2 | 920 | 130 | ||
AK352320.1 | - | 2 | 922 | 130 | ||
AK392598.1 | - | 2 | 922 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
