Assay Details
Gene Symbol: MIR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049166.1 | 1 | 44 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC106508360 | -494.1kb | Anti-sense | Chr. 13 - 125777752 - 125788021 on NCBI Build Sscrofa11.1 | |
Downstream | tumor protein p63 regulated 1 | TPRG1 | 247.7kb | Sense | Chr. 13 - 126529961 - 126860676 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
