Assay Details
Gene Symbol: MIR222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_035397.1 | 1 | 33 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-221 | MIR221 | ssc-mir-221 | -671.0 | Sense | Chr. X - 40571068 - 40571137 on NCBI Build Sscrofa11.1 |
Downstream | microRNA mir-9859 | MIR9859 | ssc-mir-9859 | 86.7kb | Anti-sense | Chr. X - 40658559 - 40658638 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
