Assay Details
Gene Symbol: MIR9859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128555.1 | 1 | 33 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-222 | MIR222 | ssc-mir-222 | -86.7kb | Anti-sense | Chr. X - 40571808 - 40571887 on NCBI Build Sscrofa11.1 |
Downstream | KRAB domain-containing protein 4-like | LOC110257782 | 431.4kb | Sense | Chr. X - 41090079 - 41116172 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
