Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356327.4 | XP_003356375.2 | 983 | 87 | ||
XM_021096010.1 | XP_020951669.1 | 659 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
