Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244806.1 | NP_001231735.1 | 1 - 2 | 194 | 130 | |
GenBank mRNA | AK343271.1 | - | 1 - 2 | 194 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
