Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244806.1 | NP_001231735.1 | 4 - 5 | 1028 | 65 | |
XM_013978601.2 | XP_013834055.2 | 2153 | 65 | |||
XM_013978602.2 | XP_013834056.2 | 2421 | 65 | |||
XM_013978603.2 | XP_013834057.2 | 1569 | 65 | |||
GenBank mRNA | AK343271.1 | - | 4 - 5 | 1028 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
