Assay Details
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256367.1 | NP_001243296.1 | 2 - 3 | 369 | 103 | |
GenBank mRNA | AF264083.1 | - | 1 - 2 | 131 | 103 | |
AY609524.1 | - | 2 - 3 | 270 | 103 | ||
JN606069.1 | - | 2 - 3 | 370 | 103 | ||
JQ026234.1 | - | 2 - 3 | 369 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
