Assay Details
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285971.1 | NP_001272900.1 | 2 - 3 | 367 | 79 | |
XM_021063998.1 | XP_020919657.1 | 215 | 79 | |||
XM_021063999.1 | XP_020919658.1 | 325 | 79 | |||
XM_021064000.1 | XP_020919659.1 | 456 | 79 | |||
GenBank mRNA | JQ012762.2 | - | 2 - 3 | 367 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
