Assay Details
Gene Symbol: FUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287420.1 | NP_001274349.1 | 12 - 13 | 1365 | 109 | |
XM_013995581.2 | XP_013851035.1 | 1460 | 109 | |||
GenBank mRNA | AK348159.1 | - | 13 - 14 | 1441 | 109 | |
JF940526.1 | - | 12 - 13 | 1365 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
