Assay Details
Gene Symbol: FUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287420.1 | NP_001274349.1 | 5 - 6 | 736 | 106 | |
XM_013995581.2 | XP_013851035.1 | 831 | 106 | |||
GenBank mRNA | AK348159.1 | - | 6 - 7 | 812 | 106 | |
JF940526.1 | - | 5 - 6 | 736 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
