Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297634.1 | NP_001284563.1 | 2 - 3 | 436 | 86 | |
XM_013996120.2 | XP_013851574.1 | 824 | 86 | |||
XM_021085693.1 | XP_020941352.1 | 863 | 86 | |||
XM_021085694.1 | XP_020941353.1 | 1141 | 86 | |||
XM_021085695.1 | XP_020941354.1 | 1233 | 86 | |||
XM_021085696.1 | XP_020941355.1 | 1217 | 86 | |||
XM_021085698.1 | XP_020941357.1 | 2622 | 86 | |||
XM_021085702.1 | XP_020941361.1 | 1141 | 86 | |||
GenBank mRNA | AK231444.1 | - | 2 - 3 | 685 | 86 | |
AY609448.1 | - | 2 - 3 | 599 | 86 | ||
KJ652909.1 | - | 2 - 3 | 436 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
