Assay Details
Gene Symbol: GPIHBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308826.1 | NP_001295755.1 | 3 - 4 | 328 | 74 | |
XM_013996411.2 | XP_013851865.1 | 363 | 74 | |||
XM_021088658.1 | XP_020944317.1 | 416 | 74 | |||
GenBank mRNA | CF175807.1 | - | 1 - 2 | 383 | 74 | |
CF177157.1 | - | 1 - 2 | 61 | 74 | ||
CF361073.1 | - | 3 - 4 | 328 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
