Assay Details
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315593.1 | NP_001302522.1 | 1 - 2 | 61 | 77 | |
XM_021080123.1 | XP_020935782.1 | 119 | 77 | |||
XM_021080130.1 | XP_020935789.1 | 40 | 77 | |||
GenBank mRNA | AK240330.1 | - | 1 - 2 | 77 | ||
KP735777.1 | - | 1 - 2 | 61 | 77 | ||
KP735784.1 | - | 1 - 2 | 61 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315593.1, XM_021080123.1, XM_021080130.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
